Please login to your online account to display your discounted pricing

Sod Pot Tartrate,0.25% (w/v), Ricca Chemical

Chemical Identifiers

Chemical Name or Material Sod Pot Tartrate
Assay Percent Range 0.25% (w/v)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

R75350001A

 
ricca chemical company
RIR75350001A
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R753500020F

 
ricca chemical company
RIR753500020F
20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R75350004A

 
ricca chemical company
RIR75350004A
4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1L
Chemical Name or Material Sod Pot Tartrate
Packaging Poly Bottle
Assay Percent Range 0.25% (w/v)