missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Wheaton™ Socorex™ Low-Profile Bottle-Top Dispensers

Semiautomatic dispensers help reduce reagent waste, and accordingly, disposal costs

Manufacturer:  DWK Life Sciences 844060

 View more versions of this product

Catalog No. 13-706-16



Includes: Wide-mouth amber reservoir (also available as accessory), delivery nozzle and plunger

Description

Description

  • Retractable, graduated column makes volume setting easy and saves space in incubators and refrigerators
  • PTFE resin coated plunger prevents the “freezing” normally associated with dispensing caustic solutions
  • Delivery nozzle locks into a threaded fitting for safe operation
  • No metal parts or springs to corrode; all parts that come in contact with the liquid are resistant to most chemicals
  • Valve is made of borosilicate glass; valve balls are of synthetic ruby
  • Syringe is made of borosilicate glass; delivery jet and unions are made of PTFE resin
  • Three models cover volume range: 0.2 to 30mL

Order Info

Can be removed for cleaning and/or autoclaving (121°C/250°F)

Specifications

Specifications

0.2 to 2mL
Yes
8.45 oz.
Low Profile Bottletop Dispenser
0.1mL
0.1mL
<0.5 to <0.1%
250mL
Wide mouth amber reservoir, delivery nozzle, plunger
1 Year
Documents

Not for use with concentrated or full-strength acids