Please login to your online account to display your discounted pricing

Soap Solution, 1mL = 1mg CaCO3, Ricca Chemical

$92.85 - $223.09

Chemical Identifiers

Chemical Name or Material Soap Solution
CAS 64-17-5
CAS Min % 74.0
CAS Max % 77.0
CAS 7732-18-5
CAS Min % 23.3
CAS Max % 24.25
CAS 8029-38-7
CAS Min % 1.17
CAS Max % 1.21
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

7110-11

 
ricca chemical company
RI7110.11
Poly natural 4L Each for $223.09

7110-132

 
ricca chemical company
RI7110.132
Poly natural 1L Each for $92.85
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 4L
Concentration or Composition (by Analyte or Components) 1mL = 1mg CaCO3
Packaging Poly natural
Chemical Name or Material Soap Solution
Shelf Life 12 months
CAS Max % 1.21
CAS 64-17-5
CAS Min % 74.0
CAS Max % 77.0
CAS 7732-18-5
CAS Min % 23.3
CAS Max % 24.25
CAS 8029-38-7
CAS Min % 1.17
Safety and Handling

12 months