Please login to your online account to display your discounted pricing

SNS 314 mesylate, Tocris™

Potent pan-Aurora kinase inhibitor

$290.95 - $1,205.80

Chemical Identifiers

Chemical Name or Material SNS 314 mesylate
Assay Percent Range >98%
Molecular Formula C18H15ClN6OS2·CH3SO3H
Formula Weight 527.04
Synonym N-(3-Chlorophenyl)-N'-[5-[2-(thieno[3, 2-d]pyrimidin-4-ylamino)ethyl]-2-thiazolyl]urea methanesulfonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

458410

 
r & d systems
RDS458410
10mg Each for $290.95

458450

 
r & d systems
RDS458450
50mg Each for $1,205.80
Description & Specifications

Specifications

Formula Weight 527.04
Quantity 10mg
Chemical Name or Material SNS 314 mesylate
Molecular Formula C18H15ClN6OS2·CH3SO3H
Recommended Storage Store at -20°C
Assay Percent Range >98%
CAS 1146618-41-8
Synonym N-(3-Chlorophenyl)-N'-[5-[2-(thieno[3, 2-d]pyrimidin-4-ylamino)ethyl]-2-thiazolyl]urea methanesulfonate
Safety and Handling

Store at -20°C