Please login to your online account to display your discounted pricing

SNOG, Tocris

NO carrier. Breaks down to release NO

$113.57 - $449.15

Chemical Identifiers

Chemical Name or Material SNOG
Assay Percent Range >98%
Molecular Formula C10H16N4O7S
Formula Weight 336.32
Synonym S-Nitrosoglutathione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

060350

 
r & d systems
RDS060350
50mg Each for $449.15

060310

 
r & d systems
RDS060310
10mg Each for $113.57
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 336.32
Chemical Name or Material SNOG
Assay Percent Range >98%
Molecular Formula C10H16N4O7S
CAS 57564-91-7
Synonym S-Nitrosoglutathione
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C