Please login to your online account to display your discounted pricing

SNOG, Tocris


NO carrier. Breaks down to release NO

Quantity 50mg

Manufacturer: r & d systems  RDS060350

 View more versions of this product

Catalog No. 060350

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 336.32
Chemical Name or Material SNOG
Assay Percent Range >98%
Molecular Formula C10H16N4O7S
CAS 57564-91-7
Synonym S-Nitrosoglutathione
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material SNOG
Assay Percent Range >98%
Molecular Formula C10H16N4O7S
Formula Weight 336.32
Synonym S-Nitrosoglutathione
Accessories

Desiccate at -20°C