missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ Human SNF1LK2, GST Tag Recombinant Protein
SNF1LK2 (QIK) belongs to the AMP-activated protein kinase family.
Supplier: Invitrogen™ PV4793
Description
SNF1LK2 (QIK) is a serine/threonine kinase and is a salt-inducible kinase.
Specifications
NP_053006.1 | |
50mM Tris pH 7.5, 150mM NaCl, 0.04% Triton X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol | |
1mg | |
RUO - research use only | |
SNF1LK2, QIK | |
Human | |
GST Tag | |
Frozen |
Nuclear Receptor Assay | |
131.4kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
CAMKL Kinase Family | |
Recombinant | |
Baculovirus |