Please login to your online account to display your discounted pricing

SNC 80, Tocris

Highly selective non-peptide δ agonist

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material SNC 80
Assay Percent Range >99%
Molecular Formula C28H39N3O2
Formula Weight 449.64
Synonym (+)-4-[(αR)-α-((2S,5R)-4-Allyl-2,5-dimethyl-1-piperazinyl)-3-methoxybenzyl]-N,N-diethylbenzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

076410

 
r & d systems
076410
10mg Each for $252.39

076450

 
r & d systems
076450
50mg Each for $1,050.42
Description & Specifications

Specifications

Chemical Name or Material SNC 80
Quantity 10mg
CAS 156727-74-1
Recommended Storage Store at Room Temperature
Assay Percent Range >99%
Molecular Formula C28H39N3O2
Formula Weight 449.64
Synonym (+)-4-[(αR)-α-((2S,5R)-4-Allyl-2,5-dimethyl-1-piperazinyl)-3-methoxybenzyl]-N,N-diethylbenzamide
Safety and Handling

Store at Room Temperature