Please login to your online account to display your discounted pricing

SNC 80, Tocris


Highly selective non-peptide δ agonist

Quantity 10mg

Manufacturer: r & d systems  076410

 View more versions of this product

Catalog No. 076410

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 449.64
Chemical Name or Material SNC 80
Synonym (+)-4-[(αR)-α-((2S,5R)-4-Allyl-2,5-dimethyl-1-piperazinyl)-3-methoxybenzyl]-N,N-diethylbenzamide
Recommended Storage Store at Room Temperature
Molecular Formula C28H39N3O2
Assay Percent Range >99%
CAS 156727-74-1

Specifications

Chemical Name or Material SNC 80
Assay Percent Range >99%
Molecular Formula C28H39N3O2
Formula Weight 449.64
Synonym (+)-4-[(αR)-α-((2S,5R)-4-Allyl-2,5-dimethyl-1-piperazinyl)-3-methoxybenzyl]-N,N-diethylbenzamide
Accessories

Store at Room Temperature