Please login to your online account to display your discounted pricing

SNAP, Tocris

A stable analog of endogenous S-nitroso compounds

$88.05 - $361.37

Chemical Identifiers

Chemical Name or Material SNAP
Assay Percent Range >98%
Molecular Formula C7H12N2O4S
Formula Weight 220.24
Synonym (S)-Nitroso-N-acetylpenicillamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

059810

 
r & d systems
RDS059810
10mg Each for $88.05

059850

 
r & d systems
RDS059850
50mg Each for $361.37
Description & Specifications

Specifications

Chemical Name or Material SNAP
CAS 79032-48-7
Recommended Storage Store at -20°C
Assay Percent Range >98%
Quantity 10mg
Molecular Formula C7H12N2O4S
Formula Weight 220.24
Synonym (S)-Nitroso-N-acetylpenicillamine
Safety and Handling

Store at -20°C