Please login to your online account to display your discounted pricing

SNAP 5089, Tocris

Subtype-selective α1A antagonist

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material SNAP 5089
Assay Percent Range >98%
Molecular Formula C36H40N4O5·HCl
Formula Weight 645.19
Synonym 5-[[[3-(4,4-Diphenyl-1-piperidinyl)propyl]amino]carbonyl]-1,4-dihydro-2,6-dimethyl-4-(4-nitrophenyl)-3-pyridinecarboxylic acid methyl ester hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

239810

 
r & d systems
RDS239810
10mg Each for $177.37

239850

 
r & d systems
RDS239850
50mg Each for $746.48
Description & Specifications

Specifications

Formula Weight 645.19
Quantity 10mg
Chemical Name or Material SNAP 5089
Assay Percent Range >98%
Synonym 5-[[[3-(4,4-Diphenyl-1-piperidinyl)propyl]amino]carbonyl]-1,4-dihydro-2,6-dimethyl-4-(4-nitrophenyl)-3-pyridinecarboxylic acid methyl ester hydrochloride
Recommended Storage Store at +4°C
Molecular Formula C36H40N4O5·HCl
CAS 157066-77-8
Safety and Handling

Store at +4°C