Please login to your online account to display your discounted pricing

Thermo Scientific™ SnakeSkin™ 10K MWCO Dialysis Tubing

Perform desalting and buffer exchange for 10 to 100mL samples with this 10,000-MW cut-off (MWCO), ready-to-use form of traditional dialysis membrane tubing.

$247.07 - $280.14

Specifications

Quantity 35 ft. (10.5m)
Formulation Stabilized cellulose membrane
MWCO 10000 Da
View More Specs
Products
Catalog Number Mfr. No. Sufficient For Price Quantity    

PI68100

 
thermo scientific™
PI68100
Approx. 19mL per 5cm length (plus 2.5cm at each end for closure) yields 100 uses Each for $247.07

PI88243

 
thermo scientific™
88243
Approx. 10mL per 5cm length (plus 2.5cm at each end for closure) yields 100 uses Each for $271.03

PI88245

 
thermo scientific™
88245
Approx. 48mL per 5cm length (plus 2.5cm at each end for closure) yields 100 uses Each for $280.14
Description & Specifications

Specifications

Quantity 35 ft. (10.5m)
Formulation Stabilized cellulose membrane
MWCO 10000 Da

Thermo Scientific SnakeSkin Dialysis Tubing, 10K MWCO, is an easy- and ready-to-use form of traditional dialysis membrane tubing that allows desalting and buffer exchange for 10 to 100mL samples.

SnakeSkin Dialysis Tubing, 10K MWCO, is composed of regenerated-cellulose and supplied as an open, pleated (telescoped) tube. SnakeSkin Dialysis Tubing is supplied in eight inch (20cm) sticks containing 35 feet of dialysis tubing having a 16, 22, or 35mm circular internal diameter (I.D.). With similar capacity to 10.5 meters of 23 to 50 diameter dry flat-width tubing, the hydrated SnakeSkin Dialysis Tubing holds approx. 2 to 10mL of sample per centimeter of length.

Highlights:

  • Convenient – ready to use, pre-wetted, pleated tube

  • High recovery – greater than 90% protein recovery

  • Speed – dialysis is generally completed in 4 to 6 hours

  • Stability – compatible with a variety of laboratory solutions, including acids, bases, hydrophobic solvents and alcohols

10K MWCO Tubing Specifications:

  • Membrane Type: Regenerated cellulose

  • Tubing Nominal Dry Thickness: 0.9 to 1.2mil (22 to 30μm)

  • Glycerol Content: 21%

  • Sulfur Content: 0.1% to 0.15%

  • Heavy Metals Content: Trace