missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Snail1 Mouse anti-Human, Alexa Fluor® 488, Clone: 20C8, eBioscience™

Mouse Monoclonal Antibody

Manufacturer:  Life Technologies LS53985980

 View more versions of this product

Catalog No. 501124656



Description

Description

Description: The monoclonal antibody 20C8 recognizes human Snail1, a 29 kDa zinc finger transcription factor. Snail1 and Slug both belong to a family of transcription factors that are responsible for inducing epithelial to mesenchymal transition (EMT) during normal organ development and tumor progression. Snail1 expression is localized to both the cytoplasm and nucleus and functions as a transcriptional repressor of E-cadherin, claudins, occludin, and desmoplakin. During embryonic development, Snail1 is localized to the mesoderm, while in the adult, expression is inducible and is usually limited to activated fibroblasts and some mesenchymal cells. Overexpression of Snail1 has been found in carcinomas and fibrosarcomas and may be used as an early marker for EMT. Applications Reported: This 20C8 antibody has been reported for use in immunohistochemical staining of formalin-fixed paraffin embedded (IHC-P) tissue, ELISA, immunoprecipitation, western blotting, immunocytochemistry, and intracellular staining followed by flow cytometric analysis. Applications Tested: This 20C8 antibody has been tested by immunocytochemistry on fixed and permeabilized MCF7 cells and by immunohistochemistry on formalin-fixed paraffin embedded human infiltrating ductal carcinoma. This antibody can be used at less than or equal to 20 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Description: The monoclonal antibody 20C8 recognizes human Snail1, a 29kDa zinc finger transcription factor. Snail1 and Slug both belong to a family of transcription factors that are responsible for inducing epithelial to mesenchymal transition (EMT) during normal organ development and tumor progression. Snail1 expression is localized to both the cytoplasm and nucleus and functions as a transcriptional repressor of E-cadherin, claudins, occludin, and desmoplakin. During embryonic development, Snail1 is localized to the mesoderm, while in the adult, expression is inducible and is usually limited to activated fibroblasts and some mesenchymal cells. Overexpression of Snail1 has been found in carcinomas and fibrosarcomas and may be used as an early marker for EMT.Applications Reported: This 20C8 antibody has been reported for use in immunohistochemical staining of formalin-fixed paraffin embedded (IHC-P) tissue, ELISA, immunoprecipitation, western blotting, immunocytochemistry, and intracellular staining followed by flow cytometric analysis.Applications Tested: This 20C8 antibody has been tested by immunocytochemistry on fixed and permeabilized MCF7 cells and by immunohistochemistry on formalin-fixed paraffin embedded human infiltrating ductal carcinoma. This antibody can be used at less than or equal to 20 ug/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Snail1
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
O95863
Zinc finger protein SNAI1, Snail protein homolog, Sna protein
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
20C8
Alexa Fluor 488
SNAI1
Liquid
SNAI1
IgG2a, kappa, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
6615
Documents

For Research Use Only.