Fisherbrand™ SMS Lab Jacket Cranberry
Disposable lab jacket with anti-static coating. Knit wrist cuffs provide comfortable and secure fit.
$15.25 - $16.19
Specifications
Color Cranberry
Closure Type Snaps
Cuff Style Knit
Collar Type Knit
Material SMS (Spunbound Melt-Blown Synthetic)
View More Specs
Products 6
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17103077 View Documents Fisherbrand™
17103077
Small Pack of 10 Each for $15.25
Case of 3 Pack for $43.77
17103078 View Documents Fisherbrand™
17103078
Medium Pack of 10 Each for $15.25
Case of 3 Pack for $43.77
17103079 View Documents Fisherbrand™
17103079
Large Pack of 10 Each for $15.25
Case of 3 Pack for $43.77
17103080 View Documents Fisherbrand™
17103080
X-Large Pack of 10 Each for $15.72
Case of 3 Pack for $45.12
17103081 View Documents Fisherbrand™
17103081
2X-Large Pack of 10 Each for $15.72
Case of 3 Pack for $45.12
17103082 View Documents Fisherbrand™
17103082
3X-Large Pack of 10 Each for $16.19
Case of 3 Pack for $46.47
Description
Description
Single-use lab coat, Made of 40gsm SMS anti-static coating:
  • One outside left breast pocket
  • Two outside lower pockets
  • Knit collar
  • Front snap closure
  • Knit cuffs
Specifications
Specifications
Cranberry
Knit
SMS (Spunbound Melt-Blown Synthetic)
3
Yes
Snaps
Knit
Unisex
Non-sterile