Fisherbrand™ SMS Lab Coats White
Disposable lab coat with anti-static coating. Knit wrist cuffs provide comfortable and secure fit.
$16.25 - $16.73
Specifications
Color White
Closure Type Snaps
Cuff Style Knit
Collar Type Knit
Material SMS (Spunbound Melt-Blown Synthetic)
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17103000 View Documents Fisherbrand™
17103000
X-Small Pack of 10 Each for $16.25
Case of 3 Pack for $46.62
17103001 View Documents Fisherbrand™
17103001
Small Pack of 10 Each for $16.25
Case of 3 Pack for $46.62
17103002 View Documents Fisherbrand™
17103002
Medium Pack of 10 Each for $16.25
Case of 3 Pack for $46.62
17103003 View Documents Fisherbrand™
17103003
Large Pack of 10 Each for $16.25
Case of 3 Pack for $46.62
17103004 View Documents Fisherbrand™
17103004
X-Large Pack of 10 Each for $16.73
Case of 3 Pack for $48.03
17103005 View Documents Fisherbrand™
17103005
2X-Large Pack of 10 Each for $16.73
Case of 3 Pack for $48.03
17103006 View Documents Fisherbrand™
17103006
3X-Large Pack of 10 Each for $16.73
Case of 3 Pack for $48.03
17103007 View Documents Fisherbrand™
17103007
5X-Large Pack of 10 Each for $16.73
Case of 3 Pack for $48.03
Description
Description
Single-use lab coat, Made of 40gsm SMS anti-static coating:
  • One outside left breast pocket
  • Two outside lower pockets
  • Knit collar
  • Front snap closure
  • Knit cuffs
Specifications
Specifications
White
Knit
SMS (Spunbound Melt-Blown Synthetic)
3
Yes
Snaps
Knit
Unisex
Non-sterile