Fisherbrand™ SMS Lab Coats Cranberry
Disposable lab coat with anti-static coating. Knit wrist cuffs provide comfortable and secure fit.
$16.25 - $16.73
Specifications
Color Cranberry
Closure Type Snaps
Cuff Style Knit
Collar Type Knit
Material SMS (Spunbound Melt-Blown Synthetic)
View More Specs
Products 7
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17103030 View Documents Fisherbrand™
17103030
Small Pack of 10 Each for $16.25
Case of 3 Pack for $46.62
17103031 View Documents Fisherbrand™
17103031
Medium Pack of 10 Each for $16.25
Case of 3 Pack for $46.62
17103032 View Documents Fisherbrand™
17103032
Large Pack of 10 Each for $16.25
Case of 3 Pack for $46.62
17103033 View Documents Fisherbrand™
17103033
X-Large Pack of 10 Each for $16.73
Case of 3 Pack for $48.03
17103034 View Documents Fisherbrand™
17103034
2X-Large Pack of 10 Each for $16.73
Case of 3 Pack for $48.03
17103035 View Documents Fisherbrand™
17103035
3X-Large Pack of 10 Each for $16.73
Case of 3 Pack for $48.03
17103036 View Documents Fisherbrand™
17103036
5X-Large Pack of 10 Each for $16.73
Case of 3 Pack for $48.03
Description
Description
Single-use lab coat, Made of 40gsm SMS anti-static coating:
  • One outside left breast pocket
  • Two outside lower pockets
  • Knit collar
  • Front snap closure
  • Knit cuffs
Specifications
Specifications
Cranberry
Knit
SMS (Spunbound Melt-Blown Synthetic)
3
Yes
Snaps
Knit
Unisex
Non-sterile