Please login to your online account to display your discounted pricing

Smopex™-105, Alfa Aesar™

For Recovery of Negatively Charged Precious Metals Complexes Used in Complex Reactions (Suzuki, Heck, Sonagashira). Recovery of Precious Metals From Catalyst Process Streams

$561.84 - $561.84

Chemical Identifiers

Chemical Name or Material Smopex-105
Name Note Weak base (pyridine functional group) anion exchanger. Exchange capacity 4-6mmol/g
Physical Form Fibers
MDL Number MFCD06202261
Synonym Vinyl pyridine grafted polyolefin fiber
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4471006

 
alfa aesar
4471006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4471014

 
alfa aesar
4471014
25g Each for $561.84

AA4471022

 
alfa aesar
4471022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Fibers
Quantity 5g
Color White
Chemical Name or Material Smopex-105
Odor Odorless
Name Note Weak base (pyridine functional group) anion exchanger. Exchange capacity 4-6mmol/g
MDL Number MFCD06202261
Synonym Vinyl pyridine grafted polyolefin fiber