Smopex™-105, Alfa Aesar™
For Recovery of Negatively Charged Precious Metals Complexes Used in Complex Reactions (Suzuki, Heck, Sonagashira). Recovery of Precious Metals From Catalyst Process Streams
Manufacturer:  Alfa Aesar 4471014
 View more versions of this product
Catalog No. AA4471014
Specifications
Specifications
Smopex-105
MFCD06202261
Fibers
Odorless
25g
Weak base (pyridine functional group) anion exchanger. Exchange capacity 4-6mmol/g
Vinyl pyridine grafted polyolefin fiber
0.3g/cm3 at 20°C
White
Documents