Please login to your online account to display your discounted pricing

Smopex-101, Alfa Aesar™

For Recovery of Precious Metals From Catalyst Process Streams, Especially Pd, Rh

Chemical Identifiers

Chemical Name or Material Smopex-101
Physical Form Fibers
MDL Number MFCD06202258
Synonym Styrene sulfonic acid grafted polyolefin fiber
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4456806

  SDS
alfa aesar
4456806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4456814

  SDS
alfa aesar
4456814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4456822

  SDS
alfa aesar
4456822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Fibers
Quantity 5g
Color Yellow
Chemical Name or Material Smopex-101
MDL Number MFCD06202258
Synonym Styrene sulfonic acid grafted polyolefin fiber
Odor Odorless