Please login to your online account to display your discounted pricing

American Educational Products Smoking Experiment Kit


Demonstrate the dangers of smoking.

Manufacturer: american educational products  100611B

 View more versions of this product

Catalog No. S52274

  • / Pack of 25 Each
 

Description & Specifications

Specifications

For Use With (Equipment) Effects of smoking kit
Includes 25 filters
Product Type Filter Refill
Age 9 to 18 years
Grade Level 4 to 12