Please login to your online account to display your discounted pricing

American Educational Products Smoking Experiment Kit


Demonstrate the dangers of smoking.

Manufacturer: american educational products  R100590

 View more versions of this product

Catalog No. S52273

  • / Each
 

Description & Specifications

Specifications

Age 9 to 18 years
For Use With (Application) Dangers of smoking
Grade Level 4 to 12
Includes 25 filters, molded plastic lid and base, aspirator bulb, color chart and instructions for five complete experiments
Product Type Smoking Experiment Kit