Please login to your online account to display your discounted pricing
American Educational Products Smoking Experiment Kit
Demonstrate the dangers of smoking.
$46.35 - $126.90
Specifications
Age 9 to 18 years
Grade Level 4 to 12
View More Specs
Includes: 
  • Filter holder
  • 25 filters
  • Squeeze bulb
  • Detailed instruction manual
Products
Catalog Number Mfr. No. Includes Product Type Price Quantity    
S52274
american educational products
100611B
25 filters Filter Refill Pack of 25 Each for $46.35
S52273
american educational products
R100590
25 filters, molded plastic lid and base, aspirator bulb, color chart and instructions for five complete experiments Smoking Experiment Kit Each for $126.90
Description & Specifications
Specifications
Age 9 to 18 years
Grade Level 4 to 12
This experiment exposes the amount and nature of the tars and nicotine produced by cigarettes. Test for the efficiency of different filters, and compare the nicotine and tar content of different brands.
  • Simple apparatus draws smoke from a lighted cigarette through a white fiberglass filter, collecting nicotine and tars on filter paper Resulting color and odor are dramatic proof of what smoking cigarettes does to the body