Thermo Scientific™ Smoke-Check Smoke Detector Challenge Particles
Check transport time of air sampling smoke detectors in cleanrooms and other critical environments.
$483.61 - $483.61
Specifications
Description Smoke Detector Challenge Particles
Quantity 10mL
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
09980571 Thermo Scientific™
SD-01
Smoke Detector Challenge Particles Each for $483.61
Specifications
Description
Check transport time of air sampling smoke detectors as required by United States NFPA Standard 72 with Thermo Scientific™ Smoke-Check Smoke Detector Challenge Particles, especially useful in cleanrooms and other critical environments.
A suspension of polystyrene particles in high purity water is nebulized using a handheld ultrasonic atomizer to mimic natural smoke, thereby eliminating residues caused by traditional oil-based sprays or real smoke.
  • Consistently triggers the smoke detector with optimized particle size and concentration
  • Will not pass through HEPA filters, ensuring the integrity of the cleanroom environment
Smoke Detector Challenge Kit:
Includes a battery-powered, portable ultrasonic nebulizer, a draft shield and a 10mL bottle of particles.
Ordering Information:
Additional bottles of Smoke-Check may be purchased separately.
Description & Specifications
Specifications
Smoke Detector Challenge Particles
10mL