Eppendorf™ SmartBlock DWP500 PROMO
Thermoblock for DeepWell Plate supports optimal temperature transfer within your sample.
Supplier:  Eppendorf™ 2231001045
Catalog No. 05414305PM
Description
Description
  • For Eppendorf™ Deepwell Plate 96/500 μL
  • Optimized block design supports optimal temperature transfer within your sample to receive ideal results
  • Eppendorf™ SmartBlock-specific calibration by integrated sensor technology maximizes temperature accuracy and homogeneity
  • Every SmartBlock is individually inspected in production for temperature accuracy and is delivered includes serial number-specific certificate for documentation purposes
  • Insulated thermoblocks for ergonomic operation so that you won't burn your fingers (PhysioCare Concept)
  • Automatic block recognition for safe handling
  • Eppendorf™ QuickRelease™ technology enables fast and simple block exchange, so no need for tools
  • Limited to ThermoMixer C and ThermoStat C
Specifications
Specifications
Deepwell Plate
Block
Block and Lid