Please login to your online account to display your discounted pricing

SMANT hydrochloride, Tocris™

Inhibits Smoothened (Smo) accumulation

$181.19 - $762.58

Chemical Identifiers

Chemical Name or Material SMANT hydrochloride
Assay Percent Range >99%
Molecular Formula C16H23BrN2O·HCl
Formula Weight 375.73
Synonym N-(4-Bromophenyl)-3, 5-dimethyl-1-piperidinepropanamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

466210

 
r & d systems
RDS466210
10mg Each for $181.19

466250

 
r & d systems
RDS466250
50mg Each for $762.58
Description & Specifications

Specifications

Formula Weight 375.73
Quantity 10mg
Chemical Name or Material SMANT hydrochloride
Assay Percent Range >99%
Molecular Formula C16H23BrN2O·HCl
CAS 1177600-74-6
Synonym N-(4-Bromophenyl)-3, 5-dimethyl-1-piperidinepropanamide hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature