Please login to your online account to display your discounted pricing

SM-21 maleate, Tocris


Presynaptic cholinergic modulator

Quantity 10mg

Manufacturer: r & d systems  075110

 View more versions of this product

Catalog No. 075110

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 453.92
Assay Percent Range >99%
Chemical Name or Material SM-21 maleate
Molecular Formula C18H24ClNO3·C4H4O4
CAS 155059-42-0
Synonym (±)-Tropanyl 2-(4-chlorophenoxy)butanoate maleate
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material SM-21 maleate
Assay Percent Range >99%
Molecular Formula C18H24ClNO3·C4H4O4
Formula Weight 453.92
Synonym (±)-Tropanyl 2-(4-chlorophenoxy)butanoate maleate
Accessories

Store at Room Temperature