Please login to your online account to display your discounted pricing

SLV 320, Tocris

Potent and selective A1 antagonist

$195.49 - $816.90

Chemical Identifiers

Chemical Name or Material SLV 320
Assay Percent Range >99%
Molecular Formula C18H20N4O
Formula Weight 308.38
Synonym trans-4-[(2-Phenyl-7H-pyrrolo[2, 3-d]pyrimidin-4-yl)amino]cyclohexanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

334410

 
r & d systems
334410
10mg Each for $195.49

334450

 
r & d systems
334450
50mg Each for $816.90
Description & Specifications

Specifications

Formula Weight 308.38
Quantity 10mg
Molecular Formula C18H20N4O
Assay Percent Range >99%
Chemical Name or Material SLV 320
CAS 251945-92-3
Synonym trans-4-[(2-Phenyl-7H-pyrrolo[2, 3-d]pyrimidin-4-yl)amino]cyclohexanol
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature