Invitrogen™ SLK Recombinant Human Protein DFS Item
SLK seems to be ubiquitously expressed.
Manufacturer:  Invitrogen™ LSPV4059
 View more versions of this product
Catalog No. PV4059
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
SLK acts as a novel effector of cytoskeletal remodeling and apoptosis.
Specifications
Specifications
NP_055535.1
Frozen
160.3kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
STE20 Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
SLK, STK2
Baculovirus
Human
GST Tag
Documents