Please login to your online account to display your discounted pricing
Thermo Scientific™ SLIPFREE™ Sample Loops
Thermo Scientific™ SLIPFREE Sample Loops feature a self-adjusting, leak-free design.
$408.71 - $617.72
Specifications
Type Sample Loop
For Use With Liquid Chromatography
View More Specs
Products
Catalog Number Mfr. No. Volume (Metric) Price Quantity    
32-650
thermo scientific™
32625
100μL Each for $617.72
32-699
thermo scientific™
32650
250μL Each for $408.71
Description & Specifications
Specifications
Type Sample Loop
For Use With Liquid Chromatography
Thermo Scientific™ SLIPFREE Sample Loops feature a self-adjusting, leak-free design.
  • Compatible with Rheodyne and Valco™ injectors
  • Long neck design