Please login to your online account to display your discounted pricing
Thermo Scientific™ SLIPFREE™ HPLC Column Double Connectors
Ensure leak-free, void-free column connections and optimize LC performance with self-adjusting Thermo Scientific™ SLIPFREE™ HPLC Column Double Connectors.
$337.04 - $337.04
Products
Catalog Number Mfr. No. Diameter (English) Inner Length (Metric) Type Price Quantity    
30211FLEX
thermo scientific™
30211FLEX
0.005 in. 10.5cm Double Flexible Each for $337.04
30215FLEX
thermo scientific™
30215FLEX
0.005 in. 10.5cm Double Flexible Pack of 4 Each for $337.04
Description & Specifications
Ensure excellent column connections and optimal HPLC performance with Thermo Scientific™ SLIPFREE™ HPLC Column Double Connectors. Compatible with all column end-fittings, these universal, self-adjusting connectors provide leak- and void-free column connections. Fingertight connections up to 10000psi ensure ease of use. They are ideal for supercritical fluid columns (SFC). SLIPFREE connectors are available in a variety of configurations and designs for a broad range of applications.
Leak-Free, Void-Free Connections:
  • Pushes tubing and ferrule into end-fitting
  • Self adjusts to fit each new column connection
  • Fingertight connection up to 10000 psi
Flexible:
  • Fits all brands of columns
  • Available in single and double configurations
  • Available in a range of internal diameters to suit multiple applications
  • PEEK-collared and long-neck designs available for specific applications