Please login to your online account to display your discounted pricing

Sling Psychrometer Kit


Determine dew point and relative humidity.

Manufacturer: fisher science education  3106

 View more versions of this product

Catalog No. S53087

  • / Each
 

Description & Specifications

Specifications

Includes 2 dry bulb thermometers, teacher guide with psychrometric tables, wet bulb wick
Product Type Sling Psychrometer Kit
For Use With (Application) To determine dew point and relative humidity
Quantity Each
Grade 6 to 12