Please login to your online account to display your discounted pricing

Sling Psychrometer Kit

Determine dew point and relative humidity.

$15.20 - $133.65

Specifications

Product Type Sling Psychrometer Kit
For Use With (Application) To determine dew point and relative humidity
Grade 6 to 12
Includes 2 dry bulb thermometers, teacher guide with psychrometric tables, wet bulb wick
View More Specs
Includes:  Wet bulb wick, two dry bulb thermometers and teacher's guide with psychrometric tables
Products
Catalog Number Mfr. No. Quantity Price Quantity    

S53087

 
fisher science education
3106
Each Each for $15.20

S53087A

 
fisher scientific
16221
15 Pk. Each for $133.65
Description & Specifications

Specifications

Product Type Sling Psychrometer Kit
For Use With (Application) To determine dew point and relative humidity
Grade 6 to 12
Includes 2 dry bulb thermometers, teacher guide with psychrometric tables, wet bulb wick

  • Temperature measured in Celsius
  • Grades 6 to 12