Please login to your online account to display your discounted pricing

Sling Psychrometer Kit


Determine dew point and relative humidity.

Manufacturer: fisher scientific  16221

 View more versions of this product

Catalog No. S53087A

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) To determine dew point and relative humidity
Grade 6 to 12
Includes 2 dry bulb thermometers, teacher guide with psychrometric tables, wet bulb wick
Quantity 15 Pk.
Product Type Sling Psychrometer Kit