Epredia™ SlideMate™ Accessories
Enhance and optimize the Epredia™ SlideMate™ Slide Printer with Epredia™ SlideMate Slide Printer Accessories.
Supplier: Epredia™ B81300031
Description
Accessories include:
- Collection system Collection trays (box of 4)
- Hopper
- Hot foil tape, box of 5 (roll of tape prints approx. 20,000 cassettes)
Specifications
Mini PC Package | |
Mini PC package with desktop stand | |
SlideMate™ Slide Printer |