Please login to your online account to display your discounted pricing
Thermo Scientific™ Slide Mailer and Folder
Hold up to 20 slides with the Thermo Scientific™ Slide Mailer and Folder.
$74.29 - $1,149.77
Products
Catalog Number Mfr. No. Material Price Quantity    
36-101-4307
thermo scientific™
2500
Cardboard Case of 24 Each for $1,149.77
58-308
thermo scientific™
58308
Plastic Each for $74.29
Description & Specifications
Hold up to 20 slides with the Thermo Scientific™ Slide Mailer and Folder.
Slide Mailer
  • Cardboard construction
  • Mailer holds two 3 × 1″ (76 × 25mm) microscope slides
  • 3.125 × 4″ (79 × 102mm) configuration conforms to U.S. Postal regulations
  • Cut-out windows allow viewing of patient identification
  • 24 per box, 30 boxes per case
Slide Folder
  • Plastic construction
  • See-through flaps eliminate need to open the folder when searching for slides
  • Easy to clean