Please login to your online account to display your discounted pricing
Thermo Scientific™ Slide and Block Cabinets
Form an integrated storage system with stackable Thermo Scientific™ Slide and Block Cabinets, constructed of steel and finished in solvent-proof paint.
$512.77 - $1,414.80
Products
Catalog Number Mfr. No. Description Color Price Quantity    
16-100-2
thermo scientific™
SH161002
Block Bank Gray Each for $1,287.02
16-100-3
thermo scientific™
161003
Block Bank Green Each for $1,287.02
16-110-0
thermo scientific™
161100
Universal Plinth Black Each for $512.77
28-618-91
thermo scientific™
SH161301
Slide Safe Blue Each for $1,414.80
16-130-2
thermo scientific™
161302
Slide Safe Gray Each for $1,030.90
16-130-3
thermo scientific™
SH161303
Slide Safe Green Each for $1,397.80
16-140-0
thermo scientific™
161400
Wheeled Plinth N/A Each for $1,404.50
Description & Specifications
Form an integrated storage system with stackable Thermo Scientific™ Slide and Block Cabinets, constructed of steel and finished in solvent-proof paint.
Features
  • Steel construction
  • Solvent-proof paint
  • Sealed top and bottom to ensure dust-free storage
  • Universal plinth in black metal for Slide Safe and Block Bank
  • Optional Wheeled Plinth capable of supporting up to ten Slide Safes
Slide Safe
  • Contains fourteen drawers
  • Total capacity of 6000 microscope slides
  • Dimensions: 18.9 × 18.9 × 5.1" (480 × 480 × 130mm)
Block Bank
  • Contains eight drawers
  • Approximate total capacity of 3000 blocks
  • Dimensions: 18.9 × 18.9 × 10.8" (480 × 480 × 275mm)
Ordering Information:
Foam dividers for slide drawers (Cat. No. B1000152) available in packs of 12.