Thermo Scientific™ Slide-A-Lyzer™ Dialysis Cassette Accessories
Load samples into Slide-A-Lyzer dialysis cassettes and float them in dialysate buffer using these syringes and buoys.
$47.66 - $85.15
Products ${productFamilyLength}
Catalog Number Mfr. No. Sufficient For Price Quantity  
Catalog Number Mfr. No. Sufficient For Price Quantity  
PI66430 Thermo Scientific™
PI66430
Floating one 0.5mL or 3mL Cassette with each buoy Pack of 10 Each for $63.78
PI66431 Thermo Scientific™
PI66431
Floating ten 0.5mL or 3mL Cassettes simultaneously Pack for $85.15
PI66432 Thermo Scientific™
PI66432
Floating one 12mL Cassette with each buoy Pack of 8 Each for $50.75
PI66490 Thermo Scientific™
PI66490
Loading samples into 3mL Cassettes Pack of 10 Each for $49.19
PI66493 Thermo Scientific™
PI66493
Loading samples into 12mL Cassettes Pack of 10 Each for $50.74
PI66494 Thermo Scientific™
PI66494
Loading samples into 0.5mL Cassettes Pack for $47.66
Specifications
Description
Thermo Scientific™ Slide-A-Lyzer™ Dialysis Cassette Accessories are syringes and buoys for loading samples and floating the cassettes in dialysate.
These accessories facilitate use of Slide-A-Lyzer Dialysis Cassettes. Three different sizes of syringes with 18-gauge beveled-tip needles enable efficient injection of samples into the various sizes of cassettes. The syringes also enable accurate and complete recovery of samples following dialysis.
Three flotation buoys enable Slide-A-Lyzer Dialysis Casettes to be suspended in buffer or other dialysate. The reusable buoys are made of low density polyethylene (LDPE). For the 0.5 and 3mL cassette sizes, single-cassette and 10-cassette buoys are offered. The 12mL cassette is wider than the other two sizes and requires its own model of buoy. The 30mL Slide-A-Lyzer Dialysis Cassettes contain their own air chamber and do not require use of a float buoy.
Highlights:
  • Reusable – polypropylene syringes with stainless steal needles and the LDPE float buoys are easily washed and reused many times
  • Complete – these are the only two kinds of special accessories one needs for trouble-free use of Slide-A-Lyzer Dialysis Cassettes
Item-Specific Features:
  • Single buoys (Part No. 66430) each hold one 0.1–0.5mL or 0.5–3mL cassette
  • Single buoys (Part No. 66432) each hold one 3–12mL capacity cassette
  • Carousel Buoys (Part No. 66431) each hold ten 0.1–0.5mL or 0.5–3mL cassettes
  • Needles supplied with the syringes are sterile, 1' long
Includes:
Each syringe pack contains 10 syringes and 10 needles.