Please login to your online account to display your discounted pricing

Thermo Scientific™ Slide-A-Lyzer™ Dialysis Cassette Accessories


Load samples into Slide-A-Lyzer dialysis cassettes and float them in dialysate buffer using these syringes and buoys.

Manufacturer: thermo scientific™  PI66431

 View more versions of this product

Catalog No. PI66431

  • / Pack
 

Description & Specifications

Specifications

Description Slide-A-Lyzer Cassette Carousel Float Buoy (for 0.5mL to 3mL cassettes)
Formulation Low density polyethylene (LDPE)
Quantity 1 buoy
Sufficient For Floating ten 0.5mL or 3mL Cassettes simultaneously

Thermo Scientific™ Slide-A-Lyzer™ Dialysis Cassette Accessories are syringes and buoys for loading samples and floating the cassettes in dialysate.

These accessories facilitate use of Slide-A-Lyzer Dialysis Cassettes. Three different sizes of syringes with 18-gauge beveled-tip needles enable efficient injection of samples into the various sizes of cassettes. The syringes also enable accurate and complete recovery of samples following dialysis.

Three flotation buoys enable Slide-A-Lyzer Dialysis Casettes to be suspended in buffer or other dialysate. The reusable buoys are made of low density polyethylene (LDPE). For the 0.5 and 3mL cassette sizes, single-cassette and 10-cassette buoys are offered. The 12mL cassette is wider than the other two sizes and requires its own model of buoy. The 30mL Slide-A-Lyzer Dialysis Cassettes contain their own air chamber and do not require use of a float buoy.

Highlights:

  • Reusable – polypropylene syringes with stainless steal needles and the LDPE float buoys are easily washed and reused many times

  • Complete – these are the only two kinds of special accessories one needs for trouble-free use of Slide-A-Lyzer Dialysis Cassettes

Item-Specific Features:

  • Single buoys (Part No. 66430) each hold one 0.1–0.5mL or 0.5–3mL cassette
  • Single buoys (Part No. 66432) each hold one 3–12mL capacity cassette
  • Carousel Buoys (Part No. 66431) each hold ten 0.1–0.5mL or 0.5–3mL cassettes
  • Needles supplied with the syringes are sterile, 1' long

Includes:

Each syringe pack contains 10 syringes and 10 needles.