missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Slide-A-Lyzer™ 7K MWCO Dialysis Cassettes

Remove buffer salts and contaminants and maximize sample recovery using this semi-permeable membrane with a 7000-dalton protein MW cut-off (MWCO).

$155.94 - $252.25

Specifications

MWCO 7000 Da
View More Specs

Disclaimers:

Slide-A-Lyzer Dialysis Cassette Technology is protected by U.S. Patent 5,503,741.



Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
PI66370 Thermo Scientific™
PI66370
Pack of 10 Each for $155.94
PI66372 Thermo Scientific™
PI66372
Each for $221.22
PI66373 Thermo Scientific™
PI66373
Pack of 10 Each for $155.94
PI66707 Thermo Scientific™
PI66707
Each for $252.25
PI66710 Thermo Scientific™
PI66710
Pack of 8 Each for $204.88
Specifications

Description

Thermo Scientific™ Slide-A-Lyzer Dialysis Cassettes (7K MWCO) facilitate simple and effective removal of buffer salts and small contaminants from proteins and other macromolecules that are larger than 7000 daltons.

Available in three different cassette sizes to accommodate sample volumes between 0.1 and 12mL, these devices are easier to use than traditional dialysis tubing, providing both faster buffer exchange and more complete sample recovery. Slide-A-Lyzer Cassettes can be used for a wide range of applications including low molecular-weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, and sample concentration.

The cassettes are constructed from two sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of a silicone-like gasket inside an inert plastic frame. The membrane and cassette materials are compatible with most common laboratory chemicals and buffers. Liquid samples are easily added and removed by penetrating the self-sealing gasket with a hypodermic needle attached to a syringe. No knots, clips or caps are needed to seal the units and prevent leaking and sample-loss during the dialysis procedure.

  • Easy to use—no knots or clamps are needed; just inject sample into cassette and begin dialysis

  • Fast dialysis—flat cassette chamber with two membranes provides high surface-area to volume ratio that maximizes diffusion rate compared to cylindrical dialysis tubing

  • High recovery—rectangular cassette design maximizes recovery of entire sample volume via any one of the four corner injection ports

  • Three cassette sizes—select the cassette that best suits your sample volume

  • Color-coded—green cassette frame specifies cassettes with the 7000 MW pore size membrane

7K Dialysis Membrane Specifications:

  • Composition: Regenerated cellulose synthesized by the Viscose method
  • Thickness: 0.9 to 1.2mil (22.5 to 30μm)
  • Hydration Required Before Use: 30 seconds for small-volume samples
  • Glycerol Content: 13%
  • Sulfur Content: 0.1 to 0.15%
  • Heavy Metals Content: Trace

Includes:

Kits contain cassettes plus an equal number of syringes with 18-gauge needles and single-cassette float buoys

Description & Specifications

Specifications

7000 Da