Please login to your online account to display your discounted pricing

Thermo Scientific™ Slide-A-Lyzer™ 3.5K MWCO Dialysis Cassettes

Remove buffer salts and contaminants and maximize sample recovery using this semi-permeable membrane with a 3500-dalton protein MW cut-off (MWCO).

$155.94 - $252.25

Specifications

MWCO 3500 Da
View More Specs
Products
Catalog Number Mfr. No. Description Price Quantity    

PI66107

 
thermo scientific™
PI66107
Slide-A-Lyzer Dialysis Cassette Kit, 3.5K MWCO, 12mL Each for $252.25

PI66110

 
thermo scientific™
PI66110
Slide-A-Lyzer Dialysis Cassettes, 3.5K MWCO, 12mL Each for $208.16

PI66130

 
thermo scientific™
66130
Slide-A-Lyzer Dialysis Cassettes, 3.5K MWCO, 30mL Pack of 6 Each for $163.27

PI66330

 
thermo scientific™
PI66330
Slide-A-Lyzer Dialysis Cassettes, 3.5K MWCO, 3mL Each for $157.58

PI66332

 
thermo scientific™
PI66332
Slide-A-Lyzer Dialysis Cassette Kit, 3.5K MWCO, 3mL Each for $221.22

PI66333

 
thermo scientific™
PI66333
Slide-A-Lyzer Dialysis Cassettes, 3.5K MWCO, 0.5mL Pack of 10 Each for $155.94

PI66335

 
thermo scientific™
PI66335
Slide-A-Lyzer Dialysis Cassette Kit, 3.5K MWCO, 0.5mL Each for $192.85
Description & Specifications

Specifications

MWCO 3500 Da

Thermo Scientific™ Slide-A-Lyzer Dialysis Cassettes (3.5K MWCO) facilitate simple and effective removal of buffer salts and small contaminants from proteins and other macromolecules that are larger than 3500 daltons.

Available in four different cassette sizes to accommodate sample volumes between 0.5 and 30mL, these devices are easier to use than traditional dialysis tubing, providing both faster buffer exchange and more complete sample recovery. Slide-A-Lyzer Cassettes can be used for a wide range of applications including low molecular-weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, and sample concentration.

The cassettes are constructed from two sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of a silicone-like gasket inside an inert plastic frame. The membrane and cassette materials are compatible with most common laboratory chemicals and buffers. Liquid samples are easily added and removed by penetrating the self-sealing gasket with a hypodermic needle attached to a syringe. No knots, clips or caps are needed to seal the units and prevent leaking and sample-loss during the dialysis procedure.

Highlights:

  • Easy to use – no knots or clamps are needed; just inject sample into cassette and begin dialysis

  • Fast dialysis – flat cassette chamber with two membranes provides high surface-area to volume ratio that maximizes diffusion rate compared to cylindrical dialysis tubing

  • High recovery – rectangular cassette design maximizes recovery of entire sample volume via any one of the four corner injection ports

  • Four cassette sizes – select the cassette that best suits your sample volume

  • Color-coded – pink cassette frame specifies cassettes with the 3500 MW pore size membrane

3.5K Dialysis Membrane Specifications:

  • Composition: Regenerated cellulose synthesized by the Viscose method
  • Thickness: 0.9 to 1.2mil (22.5 to 30μm)
  • Hydration Required Before Use: 30 seconds
  • Glycerol Content: Trace
  • Sulfur Content: 0.1 to 0.15%
  • Heavy Metals Content: Trace

Item-Specific Features:

    30mL cassette has built-in air chamber for flotation

Includes:

Kits contain cassettes plus an equal number of syringes with 18-gauge needles and single-cassette float buoys