Please login to your online account to display your discounted pricing

Thermo Scientific™ Slide-A-Lyzer™ 2K MWCO G2 Dialysis Cassettes

Remove buffer salts and contaminants while maximizing sample recovery with these second-generation self-floating protein dialysis cassettes (2K MWCO).

$152.25 - $274.88

Specifications

Formulation Cellulose membrane framed in inert plastic
MWCO 2000 Da
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) Price Quantity    

PI87717

 
thermo scientific™
87717
0.5mL Pack of 10 Each for $152.25

PI87718

 
thermo scientific™
87718
3mL Pack of 10 Each for $152.25

PI87719

 
thermo scientific™
PI87719
15mL Pack of 8 Each for $220.74

PI87721

 
thermo scientific™
87721
70mL Pack of 6 Each for $274.88
Description & Specifications

Specifications

Formulation Cellulose membrane framed in inert plastic
MWCO 2000 Da

Thermo Scientific™ Slide-A-Lyzer™ 2K MWCO G2 Dialysis Cassettes provide maximum convenience, flexibility and performance for sample dialysis.

These second generation (G2) Slide-A-Lyzer Dialysis Cassettes are free-standing, self-floating and pipette-loadable units for dialysis. The single-use, disposable cassettes are available in five different sizes to accommodated samples between 0.1mL and 70mL. They offer maximum efficiency, convenience and sample protection in one package. Sample loading and removal are easily accomplished by using a serological pipette or hypodermic needle (optional) attached to a syringe. The built-in air chamber provides sample buoyancy and vertical orientation of the cassette during dialysis.

The 2K molecular-weight cutoff (MWCO) cassettes use a cellulose membrane that is rated to completely retain proteins and other macromolecules that are larger than 2000 daltons while allowing removal of buffer salts and small contaminants. The membrane is composed of low-binding regenerated cellulose, and the cassettes are manufactured using clean-room conditions.

Highlights:

  • Easy-loading – pipet-accessible for easy sample loading and retrieval

  • Self-floating – integrated air chambers eliminate the need for float buoys

  • Sturdy construction – ensures the highest possible sample integrity and protection

  • Superior design – thoroughly researched and tested to provide fast and consistent dialysis with maximum sample recovery

  • Multiple sizes – five cassette capacities to optimally match 0.1 to 70mL sample volumes

  • Versatile – ideal for removing low molecular-weight contaminants, performing buffer exchange and desalting

2K Dialysis Membrane Specifications:

Composition: Regenerated cellulose synthesized by the Viscose method Thickness: 0.9 to 1.2mil (22.5 to 30μm) Hydration Required Before Use: 2 minutes

Glycerol Content: None

Sulfur Content: 0.17%

Heavy Metals Content: Trace