missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SLC27A4 Recombinant Protein, Abnova

$618.42 - $618.42

Specifications

Cross Reactivity Human
Format Solution
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blotting (Recombinant Protein)
Molecular Weight 51.8
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
89-012-124 Abnova Corporation
H00010999P0125
25μg Each for $618.42
Specifications

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Description & Specifications

Specifications

Human
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
51.8
8
Glutathione Sepharose 4 Fast Flow
Yes
-80°C
Solution
Antibody Production, ELISA, Protein Array, Western Blotting (Recombinant Protein)
Human SLC27A4 Full-length ORF Recombinant Protein with GST-tag at N-terminal
In vitro wheat germ expression system
12.5% SDS-PAGE stained with Coomassie Blue
Wheat Germ (in vitro)