Please login to your online account to display your discounted pricing

SL 327, Tocris

Selective inhibitor of MEK1 and MEK2; brain penetrant

$121.23 - $1,075.94

Chemical Identifiers

Chemical Name or Material SL 327
Assay Percent Range >99%
Molecular Formula C16H12F3N3S
Formula Weight 335.35
Synonym α-[Amino[(4-aminophenyl)thio]methylene]-2-(trifluoromethyl)benzeneacetonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

196910

 
r & d systems
RDS196910
10mg Each for $259.29

19691

 
r & d systems
RDS19691
1mg Each for $121.23

196950

 
r & d systems
RDS196950
50mg Each for $1,075.94
Description & Specifications

Specifications

Formula Weight 335.35
Quantity 10mg
Assay Percent Range >99%
Chemical Name or Material SL 327
Recommended Storage Desiccate at +4°C
Molecular Formula C16H12F3N3S
Synonym α-[Amino[(4-aminophenyl)thio]methylene]-2-(trifluoromethyl)benzeneacetonitrile
CAS 305350-87-2
Safety and Handling

Desiccate at +4°C