Please login to your online account to display your discounted pricing

SKPin C1, Tocris™

Inhibits Skp2-mediated p27 degradation; induces cell cycle arrest

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material SKPin C1
Assay Percent Range >98%
Molecular Formula C18H13BrN2O4S2
Formula Weight 465.34
Synonym 2-[4-Bromo-2-[[4-oxo-3-(3-pyridinylmethyl)-2-thioxo-5-thiazolidinylidene]methyl]phenoxy]acetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

481750

 
r & d systems
RDS481750
50mg Each for $976.15

481710

 
r & d systems
RDS481710
10mg Each for $239.63
Description & Specifications

Specifications

Formula Weight 465.34
Quantity 50mg
Chemical Name or Material SKPin C1
Assay Percent Range >98%
Molecular Formula C18H13BrN2O4S2
CAS 432001-69-9
Synonym 2-[4-Bromo-2-[[4-oxo-3-(3-pyridinylmethyl)-2-thioxo-5-thiazolidinylidene]methyl]phenoxy]acetic acid
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C