Please login to your online account to display your discounted pricing

SKI II, Tocris

Selective non-lipid inhibitor of sphingosine kinase

$188.60 - $794.96

Chemical Identifiers

Chemical Name or Material SKI II
Assay Percent Range >98%
Molecular Formula C15H11ClN2OS
Formula Weight 302.78
Synonym 4-[[4-(4-Chlorophenyl)-2-thiazolyl]amino]phenol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

209710

 
r & d systems
RDS209710
10mg Each for $188.60

209750

 
r & d systems
RDS209750
50mg Each for $794.96
Description & Specifications

Specifications

Formula Weight 302.78
Quantity 10mg
CAS 312636-16-1
Molecular Formula C15H11ClN2OS
Recommended Storage Store at -20°C
Synonym 4-[[4-(4-Chlorophenyl)-2-thiazolyl]amino]phenol
Assay Percent Range >98%
Chemical Name or Material SKI II
Safety and Handling

Store at -20°C