Please login to your online account to display your discounted pricing

SKF 97541, Tocris

Extremely potent GABAB agonist

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material SKF 97541
Assay Percent Range >98%
Molecular Formula C4H12NO2P
Formula Weight 137.12
Synonym 3-Aminopropyl(methyl)phosphinic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

037910

 
r & d systems
RDS037910
10mg Each for $221.01

037950

 
r & d systems
RDS037950
50mg Each for $904.70
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 137.12
Molecular Formula C4H12NO2P
CAS 127729-35-5
Synonym 3-Aminopropyl(methyl)phosphinic acid
Assay Percent Range >98%
Recommended Storage Desiccate at Room Temperature
Chemical Name or Material SKF 97541
Safety and Handling

Desiccate at Room Temperature