Please login to your online account to display your discounted pricing

SKF 96365 hydrochloride, Tocris

TRPC channel blocker; also inhibits store-operated Ca2+ entry

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material SKF 96365 hydrochloride
Assay Percent Range >99%
Molecular Formula C22H26N2O3·HCl
Formula Weight 402.92
Synonym 1-[2-(4-Methoxyphenyl)-2-[3-(4-methoxyphenyl)propoxy]ethyl-1H-imidazole hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

114710

 
r & d systems
RDS114710
10mg Each for $221.01

114750

 
r & d systems
RDS114750
50mg Each for $904.70
Description & Specifications

Specifications

Formula Weight 402.92
Quantity 10mg
Assay Percent Range >99%
Synonym 1-[2-(4-Methoxyphenyl)-2-[3-(4-methoxyphenyl)propoxy]ethyl-1H-imidazole hydrochloride
Chemical Name or Material SKF 96365 hydrochloride
Molecular Formula C22H26N2O3·HCl
CAS 130495-35-1
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature