Please login to your online account to display your discounted pricing

SKF 89976A hydrochloride, Tocris


Potent GABA uptake inhibitor; Penetrates blood brain barrier

Quantity 10mg

Manufacturer: r & d systems  RDS108110

 View more versions of this product

Catalog No. 108110

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 371.91
Quantity 10mg
Chemical Name or Material SKF 89976A hydrochloride
Molecular Formula C22H25NO2·HCl
CAS 85375-15-1
Synonym 1-(4, 4-Diphenyl-3-butenyl)-3-piperidinecarboxylic acid hydrochloride
Recommended Storage Desiccate at +4°C
Assay Percent Range >99%

Specifications

Chemical Name or Material SKF 89976A hydrochloride
Assay Percent Range >99%
Molecular Formula C22H25NO2·HCl
Formula Weight 371.91
Synonym 1-(4, 4-Diphenyl-3-butenyl)-3-piperidinecarboxylic acid hydrochloride
Accessories

Desiccate at +4°C