Please login to your online account to display your discounted pricing

SKF 86466 hydrochloride, Tocris™

Potent and selective α2 antagonist

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material SKF 86466 hydrochloride
Assay Percent Range >98%
Molecular Formula C11H14ClN·HCl
Formula Weight 232.15
Synonym 6-Chloro-N-methyl-2,3,4,5-tetrahydro-1-H-3-benzazepine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

386610

 
r & d systems
386610
10mg Each for $201.36

386650

 
r & d systems
386650
50mg Each for $844.73
Description & Specifications

Specifications

Formula Weight 232.15
Quantity 10mg
Molecular Formula C11H14ClN·HCl
CAS 86129-54-6
Recommended Storage Desiccate at Room Temperature
Chemical Name or Material SKF 86466 hydrochloride
Assay Percent Range >98%
Synonym 6-Chloro-N-methyl-2,3,4,5-tetrahydro-1-H-3-benzazepine hydrochloride
Safety and Handling

Desiccate at Room Temperature