Please login to your online account to display your discounted pricing

SKF 83566 hydrobromide, Tocris

Potent, selective D1-like antagonist

$126.32 - $530.82

Chemical Identifiers

Chemical Name or Material SKF 83566 hydrobromide
Assay Percent Range >98%
Molecular Formula C17H18BrNO·HBr
Formula Weight 413.15
Synonym 8-Bromo-2,3,4,5-tetrahydro-3-methyl-5-phenyl-1H-3-benzazepin-7-ol hydrobromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

158610

 
r & d systems
158610
10mg Each for $126.32

158650

 
r & d systems
158650
50mg Each for $530.82
Description & Specifications

Specifications

Formula Weight 413.15
Quantity 10mg
Molecular Formula C17H18BrNO·HBr
Recommended Storage Desiccate at Room Temperature
CAS 108179-91-5
Assay Percent Range >98%
Synonym 8-Bromo-2,3,4,5-tetrahydro-3-methyl-5-phenyl-1H-3-benzazepin-7-ol hydrobromide
Chemical Name or Material SKF 83566 hydrobromide
Safety and Handling

Desiccate at Room Temperature